Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylurea, 97%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
598-50-5
C2H6N2O
74.08
MFCD00007950
XGEGHDBEHXKFPX-UHFFFAOYSA-N
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
11719
CHEBI:44383
methylurea
CNC(=O)N
598-50-5
C2H6N2O
74.08
MFCD00007950
XGEGHDBEHXKFPX-UHFFFAOYSA-N
1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
11719
CHEBI:44383
methylurea
CNC(=O)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 96.0
CAS Max % 100.0
Color White
Melting Point 98.0°C to 102.0°C
Assay Percent Range 96% min. (on dry substance) (HPLC)
Infrared Spectrum Authentic
Linear Formula CH3NHCONH2
Packaging Plastic bottle
Beilstein 04, 64
Quantity 100g
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol
Formula Weight 74.08
Physical Form Crystalline Solid
Percent Purity 97%
Water 2% max.
Chemical Name or Material Methylurea, 97%
Show More Show Less

EINECSNumber : 209-935-0

RTECSNumber : YT7175000

TSCA : TSCA

RUO – Research Use Only