Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylurea, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
598-50-5
C2H6N2O
74.08
MFCD00007950
XGEGHDBEHXKFPX-UHFFFAOYSA-N
1-methylurea,n-methylurea,monomethylurea,urea, methyl,methyl urea,urea, n-methyl,methylmocovina,methylmocovina czech,methylharnstoff german,n-methyl urea
11719
CHEBI:44383
methylurea
CNC(=O)N
598-50-5
C2H6N2O
74.08
MFCD00007950
XGEGHDBEHXKFPX-UHFFFAOYSA-N
1-methylurea,n-methylurea,monomethylurea,urea, methyl,methyl urea,urea, n-methyl,methylmocovina,methylmocovina czech,methylharnstoff german,n-methyl urea
11719
CHEBI:44383
methylurea
CNC(=O)N
Color White
Melting Point 98.0°C to 102.0°C
Quantity 500g
Packaging Plastic bottle
Infrared Spectrum Authentic
Water 2% max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3NHCONH2
Beilstein 04, 64
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol
Formula Weight 74.08
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material Methylurea, 97%
Show More Show Less

EINECSNumber : 209-935-0

RTECSNumber : YT7175000

TSCA : TSCA