Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ HD Super Stainless Lab Worktable, Mobile

Moving lab equipment is now easy. Metro HD Super Stainless Lab Worktable, with an open base and built-in mobility features, is designed for cleanliness, stability, and convenience in any laboratory environment.

Manufacturer:  Metro™ MWT309FS

 View more versions of this product

Catalog No. 11-927-110


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Stainless Steel
Ideal for lab
86.4cm
96 in.
Stainless Steel
103kg
76.2cm
Mobile Table
96 x 30 x 34 in. (243.9 x 76.2 x 86.4cm)
34 in.
Four 5 in. dia. swivel casters, two with foot operated brakes
243.9cm
226 lb.
30 in.
Stainless Steel
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit