Learn More
Metro™ HD Super Stainless Lab Worktable, Mobile
Moving lab equipment is now easy. Metro HD Super Stainless Lab Worktable, with an open base and built-in mobility features, is designed for cleanliness, stability, and convenience in any laboratory environment.
$1854.00 - $3080.00
Specifications
Case Material Stainless Steel
Product Type Mobile Table
For Use With (Application) Ideal for lab
Includes Four 5 in. dia. swivel casters, two with foot operated brakes
View More Specs
Products 13
Catalog Number Mfr. No. Dimensions (L x W x H) Material Weight (Metric) Frame Type Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Material Weight (Metric) Frame Type Price Quantity & Availability  
17-987-35A
View Documents
Metro™
MWT306US
60 x 30 x 34 in. (152.4 x 76.2 x 86.4cm) Stainless Steel 63kg Open base (U-frame)
Each for $2,460.00
Add to cart
91-158
View Documents
Metro™
MWT307US
72 x 30 x 34 in. (182.5 x 76.2 x 86.4cm) Stainless Steel 71kg Open base (U-frame)
Each for $3,080.00
Add to cart
11-926-224
View Documents
Metro™
MWT307FS
72 x 30 x 34 in. (182.5 x 76.2 x 86.4cm) Stainless Steel 82.5kg With Undershelf
Each for $2,804.00
Add to cart
11-927-102
View Documents
Metro™
MWT305FS
48 x 30 x 34 in. (121.9 x 76.2 x 86.4cm) Stainless Steel 47.6kg With Undershelf
Each for $2,111.00
Add to cart
11-927-103
View Documents
Metro™
MWT305US
48 x 30 x 34 in. (121.9 x 76.2 x 86.4cm) Stainless Steel 58.9kg Open base (U-frame)
Each for $1,854.00
Add to cart
11-927-106
View Documents
Metro™
MWT306HS
60 x 30 x 34 in. (152.4 x 76.2 x 86.4cm) Stainless Steel 56.2kg Open base (H-frame)
Each for $2,460.00
Add to cart
11-927-105
View Documents
Metro™
MWT306FS
60 x 30 x 34 in. (152.4 x 76.2 x 86.4cm) Stainless Steel 72.5kg With Undershelf
Each for $2,294.00
Add to cart
11-927-108
View Documents
Metro™
MWT307HS
72 x 30 x 34 in. (182.5 x 76.2 x 86.4cm) Stainless Steel 70.7kg Open base (H-frame)
Each for $3,080.00
Add to cart
11-927-111
View Documents
Metro™
MWT309HS
96 x 30 x 34 in. (243.9 x 76.2 x 86.4cm) Stainless Steel 92kg Open base (H-frame)
Each for $2,336.00
Add to cart
11-927-110
View Documents
Metro™
MWT309FS
96 x 30 x 34 in. (243.9 x 76.2 x 86.4cm) Stainless Steel 103kg With Undershelf
Each for $2,680.00
Add to cart
11-927-112
View Documents
Metro™
MWT309US
96 x 30 x 34 in. (243.9 x 76.2 x 86.4cm) Stainless Steel 91kg Open base (U-frame)
Each for $2,343.00
Add to cart
11-927-104
View Documents
Metro™
MWT306FC
60 x 30 x 34 in. (152.4 x 76.2 x 86.4cm) Chrome 66.2kg With Undershelf This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-927-107
View Documents
Metro™
MWT307FC
72 x 30 x 34 in. (182.5 x 76.2 x 86.4cm) Chrome 81.6kg With Undershelf This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • With four 5 in. diameter (12.7cm) swivel casters, two with foot-operated brakes
  • Seamless, 14-gauge stainless-steel work surface with rounded edges
  • Comfortable 34 in. (86.4cm) height
  • Support structure virtually eliminates “drumming”
Order Info
Shipped knocked down. Assembles easily without tools. Casters sold separately.
Specifications
Specifications
Stainless Steel
Mobile Table
Ideal for lab
Four 5 in. dia. swivel casters, two with foot operated brakes