Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ HD Super Stainless Lab Worktable, Stationary Available on GSA/VA Contract for Federal Government customers only.

Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.

Manufacturer:  Metro™ WT306US

 View more versions of this product

Catalog No. 91-152


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Open with side and rear crossrails
30 x 30 x 34 in. (76.2 x 76.2 x 86.36cm)
34 in.
Bottom 3-sided frame (SS)
76.2cm
Stationary workTable
30 in.
76.2cm
NSF
Ideal for lab
86.36cm
30 in.
Stainless Steel
115 lb.
52kg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit