Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ HD Super Stainless Laboratory Worktable Accessory, Utility Hooks

Keep your workspace organized and items in easy reach. The Metro Utility Hooks are perfect for use with HD Super Stainless Laboratory Worktables.

Manufacturer:  Metro™ HK23C

 View more versions of this product

Catalog No. 19-109-102


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

NSF
4.29cm
Cantilever shelves for worktables with overhead
8.7cm
Snap-on
5cm
1.69 in.
Chrome-plated
3.44 in.
S
2 in.
Utility hook
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit