Metro™ MetroMax i Basket Glassware Cart

Store or move your glassware around the lab with confidence. MetroMax I Basket Glassware Cart is a Level 2 cart with built-in Microban antimicrobial product protection that allows you to configure its arrangement, ensuring storage and movement won't disrupt your workday.

Manufacturer: Metro™ GWBX32

 View more versions of this product

Catalog No. 13-451-092

  Check Availability
Add to Cart

    Specifications

    Design 4-Basket Unit
    For Use With (Application) Glassware transportation
    Includes Solid MetroMax i bottom shelf
    Height (English) 60 in.
    Height (Metric) 15.2cm
    Length (English) 36 in.
    Length (Metric) 91.4cm
    Width (English) 18 in.
    Product Type Glassware cart
    Width (Metric) 45.7cm
    Dimensions (L x W x H) 36 x 18 x 6 in. (91.4 x 45.7 x 15.2cm)
    Load Bearing Capacity (English) 800 lb.
    Load Bearing Capacity (Metric) 362.8kg
    Material Polymer

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit