Learn More
Metro™ MetroMax i™ Lab Worktable, Stainless Top and Backsplash with Solid i Shelf Available on GSA/VA Contract for Federal Government customers only.
The Metro MetroMax i™ Lab Worktable advanced polymer construction provides excellent corrosion-resistance performance for the most corrosive lab environments.
Manufacturer:  Metro™ LT36XS3
 View more versions of this product
Catalog No. 11-100-208
Add to cart
Description
Description
  • Table features smooth surfaces that are easy to clean
  • Mobile units can be easily moved to clean behind table area
  • Available top materials include Type 304 stainless and TRESPA phenolic resin (black or gray)
  • Wide range of accessories and caster options available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Casters must be ordered separately.
Specifications
Specifications
Stainless Steel
Ideal for lab
100.3cm
35.75 in.
600 lb.
Polymer
30 in.
35.75 x 30 x 39.5 in. (90.8 x 76.2 x 100.3cm)
39.5 in.
Backsplash with solid Metro™ MetroMax i™ shelf
90.8cm
273kg
Stationary Table
76.2cm