Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax i™ Polymer Lab Worktable, Black Phenolic Top with Solid i Shelf

Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.

Manufacturer:  Metro™ LT30XPB3

 View more versions of this product

Catalog No. 11-100-200


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Phenolic
Ideal for lab
101.6cm
29.75 in.
600 lb.
Polymer
29.75L x 30W x 40.5 in.H (74.9 x 76.2 x 101.6cm)
76.2cm
29.75 x 30 x 40.5 in. (74.9 x 76.2 x 101.6cm)
40.5 in.
Solid Metro™ MetroMax i™ shelf
74.9cm
273kg
Stationary Table
30 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit