Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Gray Phenolic Top with 3-Sided Frame
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Manufacturer:  Metro™ LT36XUPG3
 View more versions of this product
Catalog No. 11-100-210
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Phenolic
Ideal for lab
101.6cm
35.75 in.
600 lb.
Polymer
35.75L x 30W x 40.5 in.H (90.8 x 76.2 x 101.6cm)
76.2cm
35.75 x 30 x 40.5 in. (90.8 x 76.2 x 101.6cm)
40.5 in.
3 Sided frame
90.8cm
273kg
Stationary Table
30 in.