Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Gray Phenolic Top with Solid i Shelf
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Manufacturer:  Metro™ LTM48XPG3
 View more versions of this product
Catalog No. 11-100-237
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Phenolic
Ideal for lab
101.6cm
47.75 in.
600 lb.
Polymer
47.75L x 30W x 40.5 in.H (121.3 x 76.2 x 101.6cm)
76.2cm
47.75 x 30 x 40.5 in. (121.3 x 76.2 x 101.6cm)
40.5 in.
Solid Metro™ MetroMax i™ shelf, adaptor insert
121.3cm
273kg
Mobile Table
30 in.