Learn More
Metro™ MetroMax i™ Polymer Lab Worktable, Gray Phenolic Top with Solid i Shelf Available on GSA/VA Contract for Federal Government customers only.
Keep your lab cleaner with the Metro MetroMax i™ Polymer Lab Worktable, featuring superior corrosion resistance with built-in Microban™ antimicrobial product protection.
Manufacturer:  Metro™ LTM60XPG3
 View more versions of this product
Catalog No. 11-100-243
Add to cart
Description
Description
These worktables provide the ideal product platform, are modular by design and offer ultimate flexibility based on the application requirements.
  • Advanced polymer construction for corrosive lab environments
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Casters must be ordered separately.
Specifications
Specifications
Phenolic
Ideal for lab
101.6cm
59.75 in.
600 lb.
Polymer
59.75L x 30W x 40.5 in.H (151.7 x 76.2 x 101.6cm)
76.2cm
59.75 x 30 x 40.5 in. (151.7 x 76.2 x 101.6cm)
40.5 in.
Solid Metro™ MetroMax i™ shelf, adaptor insert
151.7cm
273kg
Mobile Table
30 in.