Metro™ MW Series Utility Cart with Wire Shelves

Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Wire Shelves make a smart addition to any workplace.

Manufacturer: Metro™ MW702

 View more versions of this product

Catalog No. 19-138-771

  Check Availability
Add to Cart

    Specifications

    Height (English) 38 in.
    Height (Metric) 97cm
    Length (English) 24 in.
    Length (Metric) 61cm
    Width (English) 18 in.
    Product Type Utility cart
    Width (Metric) 46cm
    Dimensions (L x W x H) 24 x 18 x 38 in. (61 x 46 x 97cm)
    Load Bearing Capacity (English) 375 lb.
    Load Bearing Capacity (Metric) 170kg
    Design 3-tier Model
    No. of Shelves 3
    For Use With (Application) Transporting materials that require an expanse of flat, solid surface
    Includes Super Erecta™ 3 Stainless Steel wire shelves with Plastic split sleeves, 2 one-piece Stainless Steel Handle, 4 in. dia. caster, 3.5 in. donut bumpers
    Certifications/Compliance NSF
    Handle Material Stainless steel
    Shelf Material Stainless steel

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit