Learn More
Metro™ Super Adjustable Super Erecta™ Wire Shelf, Metroseal 3 Finish Available on GSA/VA Contract for Federal Government customers only.
Customize your lab storage to suit your requirements. Metro's Super Adjustable Super Erecta™ Wire Shelf features Metroseal 3, a green epoxy finish with built-in Microbahn™ antimicrobial protection plus a unique open-wire design that permits free circulation of air and minimizes dust accumulation.
Manufacturer:  Metro™ A1824NK3
 View more versions of this product
Catalog No. 19-037463A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
24 x 18 in. (61 x 45.7cm)
Four non conductive sleeve/wedges
61cm
18 in.
Metro™ Super Erecta™ adjustable shelving system
24 in.
Metroseal
45.7cm