Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Metro™ Super Adjustable™ Super Erecta™ Wire Shelf - Stainless Steel Finish
Design your storage to suit your lab's needs. Metro Super Adjustable Super Erecta™ Wire Shelf in stainless steel has a unique open-wire design permiting free circulation of air and minimizes dust accumulation.
Manufacturer: Metro™ A3672NS
 View more versions of this product
Catalog No. 19-030-925
  N/A
Add to Cart
Includes: One pack of nonconductive wedges (four pairs).
Required Accessories : SiteSelect™ Posts (sold separately).
Related Products
Product Content Feedback