Learn More
Metro™ Super Erecta™ Swedged Posts
For cart-washable and autoclave applications
$82.00 - $99.00
Specifications
Material Stainless Steel
For Use With (Application) Ideal for autoclavable and cart washable application
For Use With (Equipment) Metro™ Super Erecta™ stem casters
View More Specs
Products 3
Catalog Number Mfr. No. Height (English) Height (Metric) Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Height (Metric) Length (English) Length (Metric) Price Quantity & Availability  
13-451-072
View Documents
Metro™
33UPSSW
33 in. 83.82cm 33 in. 87.5cm Each for $82.00
Add to cart
13-451-073
View Documents
Metro™
54UPSSW
54 in. 137.1cm 54 in. 137cm Each for $92.00
Add to cart
13-451-074
View Documents
Metro™
63UPSSW
63 in. 160cm 63 in. 160cm Each for $99.00
Add to cart
Description
Description
  • Each Type 304 Stainless Steel post has an aluminum cap swedged into the top
  • For use with standard stem casters
Specifications
Specifications
Stainless Steel
Ideal for autoclavable and cart washable application
Metro™ Super Erecta™ stem casters