Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
Supplier: Metro™ TTM24C
Includes: Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
Order Info
Shelves are ordered separately. One kit required per mobile unit.
Specifications
Mobile Unit Kit | |
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves | |
Chrome | |
15.3 kg | |
26.25 in. | |
Steel |
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system | |
61 cm | |
34 lb. | |
24 in. | |
66.7 cm |