Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ Super Erecta™ Wire Shelf, Chrome Finish

Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in chrome finish allow adjustable configurations when used with Metro Super Erecta shelving systems

Manufacturer:  Metro™ 3636NC

 View more versions of this product

Catalog No. 19-030-968


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

800 lb., 363kg
Metro™ Super Erecta™ shelving system
36 in.
36 in.
800 lb.
Chrome
36 x 36 in. (91 x 91cm)
Plastic sleeves and wedges
91cm
91cm
363kg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit