Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Configure your lab storage as needed. Metro Super Erecta Wire Shelves are easily adjustable and the Metroseal 3 green epoxy with Microban finish means they are easy to keep clean. Shelves are for use with Metro Super Erecta shelving systems.
Supplier: Metro™ 2142NK3
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Corrosion resistant finish for wet or high humidity environments
- Antimicrobial is built into finish to keep product clean
Specifications
21 x 42 in. | |
Metroseal 3 with Microban |