Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems
Supplier: Metro™ 1430NS
Description
- Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
14 in. | |
35.5 x 76 in. (14 x 30 cm) | |
Plastic sleeves and wedges |
355 mm | |
Metro™ Super Erecta™ shelving system | |
Stainless Steel |