Learn More
Metro™ Super Erecta™ Wire Shelving Accessory, Stem Casters
Choose from various types of casters to match your corrosion resistance needs
Manufacturer:  Metro™ 5MDBGSA
 View more versions of this product
Catalog No. 13-450-952
  Check Availability
Add to cart
Alert:
The total weight of equipment and load should not exceed three times the load rating per caster.
Specifications Description & Specifications
Specifications
No
150 lb.
12.7cm
Stainless Steel
-40 to 180°F
High Modulus Donut
68kg
5 in.
Metro™ Super Erecta™ wire shelving
Stainless Steel Cart-washable Brake Stem Caster
-40 to 82°C
Yes