Learn More
Metro™ Type 304 Stainless Steel Garment Racks
Open wire construction promotes laminar air flow, Metro Type 304 Stainless Steel Garment Racks have unique hanger capture mechanism that provides theft deterrance.
$1801.00 - $2681.00
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
17-988-160
![]() |
Metro™
GRW2436S |
36 in. | 91.4 cm |
Each for $1,801.00
|
|
17-988-162
![]() |
Metro™
GRW2460S |
60 in. | 152.4 cm |
Each for $2,471.00
|
|
17-988-161
![]() |
Metro™
GRW2448S |
48 in. | 122 cm |
Each for $1,945.00
|
|
17-988-163
|
Metro™
GRW2472S |
72 in. | 182.5 cm |
Each for $2,681.00
|
|
Description
- Type 304 Stainless steel and Proven Super Erecta™ construction throughout
- Floor-mountable upright and wall mount versions available
- Open wire construction promotes laminar air flow
- Electro-polished, Type 304 Stainless steel hangers are included. Hangers are spaced at 7.62cm (3 in.) intervals
- Unique hanger capture mechanism provides theft deterrance
- Easy to assemble
- Adjustable stainless steel triangular foot plates provided for floor mounting
- Upright unit is non-conductive, but can be made conductive with the addition of aluminum split sleeves (9986Z)
- Floor Mount style is 189.5cm (74.625 in.) tall. Wall Mount style is 19.05cm (7.5 in.) high
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
24 in. | |
61 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.