Metrohm 876 Manual Titrator Plus
Manual titrator with result calculation for titration with color indicator.
Manufacturer: Metrohm 28760110
 View more versions of this product
Catalog No. 02-075-1226
  Check Availability
Add to Cart
Includes: Stirring bars (16mm/25mm), support stand, clamping ring, 400mm support rod, electrode holder, manual dosing controller, USB MINI - USB A adapter, hex keys (2mm/5mm), wrench and exchange unit (20mL)
    Specifications
    Includes Stirring bars (16mm/25mm), support stand, clamping ring, 400mm support rod, electrode holder, manual dosing controller, USB MINI - USB A adapter, hex keys (2mm/5mm), wrench and exchange unit (20mL)
    For Use With Titrino/Titrino plus titrators
    Model 876 Manual Titrator Plus
    • Manual titrator with result calculation for manual titrations and dosings
    • With intelligent exchange unit (20mL), manual dosing controller and 801 magnetic stirrer including stand
    • Dialogue languages: DE, EN, ES, FR
    • Increments per cylinder volume: 10’000
    • With printer connection (USB), remote interface, RS232 connection for balance, remote control via RS232 (method load-up, start, stop) and pulse mode
    Dispensing Modes
    Manual Dosing (DOS):Push-button dosing; Particularly suitable for carrying out manual titrations with a color indicator; From the added volume the results can be calculated automatically and the report printed out; Different calculation parameters can be defined beforehand
    Extended Dosing (XDOS):Adding a fixed volume: enter volume and dosing rate; Time-controlled dosing: enter volume and time; Constant dosing rate: enter dosing rate and time
    Manual titration
    Tandem dosing
  • Description & Specifications
Related Products
Product Content Feedback