Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax Mobile Security Shelving Unit

Protect valuable materials and sensitive items from loss or pilferage with MetroMax Mobile Security Shelving Units.

$3290.00 - $4030.00

Specifications

Autoclavable Not Autoclavable
Color Taupe
Material Epoxy-coated Steel
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
For Use With (Equipment) MetroMax Shelving
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
13-460-395
View Documents
Metro™
MQSEC53VEPL
40.75 in.
Each for $3,290.00
Add to cart
 
13-460-393
View Documents
Metro™
MQSEC55VEPL
52.75 in.
Each for $3,810.00
Add to cart
 
13-460-394
View Documents
Metro™
MQSEC56VEPL
64.75 in.
Each for $4,030.00
Add to cart
 
Description

Description

  • Allows up to 20 different codes and has a 5+ year battery life. (2) AAA batteries included.
  • Double door with ergonomic 1/4-turn handle.
  • Each door opens 270 degrees and can be secured along the sides of the unit.
  • Shipped knocked down to save on freight costs.
  • Assembles in minutes.
  • Includes four corrosion resistant 5 in. diameter casters (two swivel and two swivel with brakes).
  • Add an optional intermediate shelf for increased storage capabilities.
Specifications

Specifications

Not Autoclavable
Epoxy-coated Steel
MetroMax Shelving
2
Yes
Taupe
All Laboratory and Clean Room Non-Autoclavable Applications
67.8 in.
27.81 in.
Electronic PIN Lock
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit