Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro™ MetroMax Stationary Security Shelving Unit

MetroMax Stationary Security Shelving Units keep valuable items safe at your fingertips.

$2790.00 - $3540.00

Specifications

Autoclavable Not Autoclavable
Color Taupe
Material Epoxy-coated Steel
For Use With (Application) All Laboratory and Clean Room Non-Autoclavable Applications
For Use With (Equipment) MetroMax Shelving
View More Specs

Products 3
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
13-460-301
View Documents
Metro™
MQSEC53EPL
38.875 in.
Each for $2,790.00
Add to cart
 
13-460-300
View Documents
Metro™
MQSEC55EPL
50.875 in.
Each for $3,310.00
Add to cart
 
13-460-302
View Documents
Metro™
MQSEC56EPL
62.875 in.
Each for $3,540.00
Add to cart
 
Description

Description

  • Allows up to 20 different codes and has a 5+ year battery life
  • (2) AAA batteries included.
  • Heavy duty open wire construction keeps contents visible at all times, making it easy to check inventory.
  • Double door with ergonomic 1/4-turn handle.
  • Each door opens 270 degrees and can be secured along the sides of the unit.
  • Shipped knocked down to save on freight costs.
  • Assembles in minutes.
  • Add an optional intermediate shelf for increased storage capabilities.
  • Compatible with all MetroMax i, 4 and Q shelves.
Specifications

Specifications

Not Autoclavable
Epoxy-coated Steel
MetroMax Shelving
2
No
Taupe
All Laboratory and Clean Room Non-Autoclavable Applications
66 in.
26.94 in.
Electronic PIN Lock
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit