Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Metronidazole, 50mg/mL in 0.1N HCl, sterile-filtered

Click to view available options
Quantity:
25mL
50mL
443-48-1
C6H9N3O3
171.156
MFCD00009750
VAOCPAMSLUNLGC-UHFFFAOYSA-N
4173
CHEBI:6909
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
CC1=NC=C(N1CCO)[N+](=O)[O-]
443-48-1
C6H9N3O3
171.156
MFCD00009750
VAOCPAMSLUNLGC-UHFFFAOYSA-N
4173
CHEBI:6909
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
CC1=NC=C(N1CCO)[N+](=O)[O-]
Physical Form Liquid
Quantity 50mL
Chemical Name or Material Metronidazole, 50mg/mL in 0.1N HCl, sterile-filtered

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NI5600000

storageNote1 : -30° to -10°C