Learn More
Aviva Systems Biology Corp METT10D Antibody - N-terminal region (ARP47674_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47674_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against METT10D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human METT10D. Alternate names: MGC3329, METT10DApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59631