Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ METT10D Monoclonal Antibody (OTI3B5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525956

Catalog No. PIMA525956


Only null left
Add to Cart

Description

Description

METT10D Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

METT10D is a probable methyltransferase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

METT10D
Monoclonal
1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q86W50
METTL16
Full length human recombinant protein of METT10D produced in HEK293T cell.
100 μL
Primary
Human, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3B5
Unconjugated
METTL16
2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase
Mouse
Affinity Chromatography
RUO
79066
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.