Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METT10D Mouse anti-Human, Clone: OTI3E8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504665

Catalog No. 50-167-5306


Add to cart

Description

Description

METT10D is a probable methyltransferase.
Specifications

Specifications

METT10D
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q86W50
METT10D
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79066
Immunohistochemistry (Paraffin), Western Blot
OTI3E8
Unconjugated
METTL16
Liquid
METTL16
Full length human recombit protein of human METT10D produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit