Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METT10D Mouse anti-Human, Mouse, Canine, DyLight 550, Clone: OTI3E8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272653R
Description
METT10D Monoclonal specifically detects METT10D in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
METT10D | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
METTL16 | |
Full length recombinant protein of human METT10D (NP_076991) produced in HEK293T cells. | |
0.1 mL | |
79066.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3E8 | |
Western Blot, Immunohistochemistry | |
EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title