Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METT10D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA523518
Description
METT10D Polyclonal Antibody for Western Blot, IHC (P)
METT10D is a probable methyltransferase.
Specifications
METT10D | |
Polyclonal | |
Unconjugated | |
METTL16 | |
2610100D03Rik; 2810013M15Rik; A830095F14Rik; AI846133; methyltransferase 10 domain containing; methyltransferase 10 domain-containing protein; methyltransferase like 16; methyltransferase-like protein 16; METT10D; METTL16; N6-adenosine-methyltransferase METTL16; putative methyltransferase METT10D; RNA N6-adenosine-methyltransferase METTL16; U6 small nuclear RNA (adenine-(43)-N(6))-methyltransferase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
67493, 79066 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q86W50, Q9CQG2 | |
METTL16 | |
KLH conjugated synthetic peptide between 444-472 amino acids from the C-terminal region of human METTL16. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction