Learn More
METTL1 purified MaxPab mouse Mouse anti-Human, Polyclonal, Abnova™
Mouse Polyclonal Antibody
Manufacturer:  Abnova Corporation H00004234B01P
Catalog No. 89-032-201
Add to cart
Description
Description
Mouse polyclonal antibody raised against a full-length human METTL1 protein.
  • High specificity
  • High reactivity
Specifications
Specifications
METTL1 purified MaxPab mouse
Unconjugated
In 1x PBS, pH 7.4
Liquid
METTL1
METTL1 (AAH00550, 1 a.a. ~ 276 a.a) full-length human protein.
RUO
Primary
4234
Western Blot
Mouse polyclonal antibody raised against a full-length human METTL1 protein.
BC000550
C12orf1|FLJ95748|TRM8|YDL201w
Mouse
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)