Learn More
METTL16 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304191AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
METT10D is a probable methyltransferase.Specifications
METTL16 | |
Polyclonal | |
Unconjugated | |
METTL16 | |
METT10D, methyltransferase 10 domain containing, methyltransferase 10 domain-containing protein, methyltransferase-like protein 16, putative methyltransferase METT10D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79066 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86W50 | |
METTL16 | |
Between 250 and 300 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.