Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL19 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | METTL19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170636
|
Novus Biologicals
NBP170636 |
100 μL |
Each of 1 for $436.00
|
|
Description
METTL19 Polyclonal specifically detects METTL19 in Human samples. It is validated for Western Blot.Specifications
METTL19 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
152992 | |
Synthetic peptides corresponding to C4ORF23 The peptide sequence was selected from the N terminal of C4ORF23. Peptide sequence LTPWIPVIAARSSYNCRFFVLPCCFFDFIGRYSRRQSKKTQYREYLDFIK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ12891, FLJ35725, methyltransferase like 19, methyltransferase-like protein 19, probable tRNA (uracil-O(2)-)-methyltransferase | |
TRMT44 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title