Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ METTL21C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546531
Description
Peptide sequence: AQQPGRRGEG LSSPGGWLEA EKKGAPQKDS TGGVLEESNK IEPSLHSLQK Sequence homology: Human: 100%.
METTL21C is a protein-lysine methyltransferase.
Specifications
METTL21C | |
Polyclonal | |
Unconjugated | |
Mettl21c | |
A530098C11Rik; C13orf39; methyltransferase like 21C; methyltransferase-like protein 21C; METTL21C; Protein-lysine methyltransferase METTL21C; RGD1561425 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
196541 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5VZV1 | |
Mettl21c | |
Synthetic peptide directed towards the N-terminal of human METTL21C (aa 8-57). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction