Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL25 Mouse anti-Human, Biotin, Clone: OTI2B6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272656B
Description
METTL25 Monoclonal specifically detects METTL25 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
METTL25 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
METTL25 | |
Full length recombinant protein of human METTL25 (NP_115606) produced in HEK293T cells. | |
0.1 mL | |
84190.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2B6 | |
Western Blot, Immunohistochemistry | |
C12orf26, chromosome 12 open reading frame 26, FLJ22789, hypothetical protein LOC84190 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title