Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL2A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen METTL2A
Applications Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB096073
View Documents
Novus Biologicals
NBP304709100UL
100 μg Each for $320.71
Add to cart
 
NB096074
View Documents
Novus Biologicals
NBP30470920UL
20 μg Each for $138.37
Add to cart
 
Description

Description

METTL2A Polyclonal antibody specifically detects METTL2A in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

METTL2A
Unconjugated
Rabbit
Human
FLJ12760, HSPC266, methyltransferase like 2A, methyltransferase-like protein 2A, METTL2
Recombinant fusion protein containing a sequence corresponding to amino acids 139-378 of human METTL2A (NP_859076.3).
Polyclonal
Affinity purified
Immunofluorescence
Purified
RUO
PBS with 50% glycerol, pH7.3.
339175
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit