Learn More
METTL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177181AP
Catalog No. 50-173-1740
Add to cart
Description
Description
METTL4 gene ontology annotations related to this gene include methyltransferase activity; nucleic acid binding.
Specifications
Specifications
METTL4
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U034, Q8N3J2
HsT661, methyltransferase like 4, METTL4
Rabbit
IgG
150 μL
-20°C
Primary
316731, 64863, 76781
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
METTL4
Liquid
Mettl4
METTL4 Fusion Protein Ag12093
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat