Learn More
Abnova™ METTL7A 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00025840T01
Catalog No. 89-952-240
Add to cart
Description
Description
Specifications
Specifications
293T
NM_014033
pCMV-METTL7A full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
METTL7A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 28.3kDa
Transient overexpression cell lysate was tested with anti-METTL7A antibody (Mfr. No. H00025840-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human