Learn More
Mettler Toledo™ Analytical Balances Density Determination Kits for AB, AB-S, and PB Series Balances
For the density determination of solids.
Manufacturer:  Mettler Toledo™ 33360
Catalog No. 10-820-14
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 250mL beaker, thermometer, bridge, gem holder, two brackets, and 25mL bottle of wetting agent.
Description
Description
  • For use with AB-S/PB-S Solids/AB-S (PB 1mg models only)
Specifications
Specifications
AB, AB-S and PB series balance
Determining densities of solids
250mL Beaker, thermometer, bridge, gem holder, two