Learn More
Mettler Toledo™ Antitheft Devices
For discontinued AB, PR, PB, SR, PG-S, SB, College B and BD Series Balances.
Manufacturer:  Mettler Toledo™ 590101
Catalog No. 01-910-52
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Cable device consists of cable lock with key. Tube device consists of aluminum tube, bar, pin, and screws. Screw aluminum tube to back of balance and insert bar. Use bar to firmly attach pin to the balance. Thread pin through a hole in balance table and secure balance with standard padlock (not included).
Cable Device
  • Consists of cable lock with key
  • Tube Device
    • Consists of aluminum tube, bar, pin and screws
    • Simple to use: Screw aluminum tube to back of balance and insert bar; use bar to firmly attach pin to the balance; thread pin through a hole in balance table and secure balance with standard padlock (not included)
    Specifications
    Specifications
    Anti-theft Device
    Steel
    60cm
    AB, PR, PB, SR, PG-S, SB, college B and BD series
    23.6 in.