Learn More
Mettler Toledo™ CarePacs™: Customized Third Weight
Third weight for customer-specific testing (e.g., minimum weight determination)
Manufacturer:  Mettler Toledo™ 11123151
 View more versions of this product
Catalog No. 01-912-123
Add to cart
Description
Description
  • Optional customized third weight for Mettler Toledo CarePacs
  • ASTM or OIML certified
Specifications
Specifications
Customized 3rd weight
ASTM Class 1
200mg
Each
ISO9001, ISO14001 environmental standard
Stainless Steel
200mg
1