Learn More
Mettler Toledo Classic Light Toploading Balances
Designed for essential weighing operations
Includes: AC adapter appropriate to national standard
Catalog Number | Mfr. No. | Model | Capacity | Readability | Stabilization Time | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Capacity | Readability | Stabilization Time | Price | Quantity & Availability | ||
01-910-160
|
Mettler Toledo
12110043 |
PL602-S/03 | 610 g | 0.01 g | 2.5 sec. |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
For simple, easy, weighing tasks that need to be done frequently, but with assured accuracy: piece counting, percent weighing, ±weighing, dynamic weighing, and calculation with a custom “free factor”
Key Features
- Logical menu guidance with only a few keys for full function
- Portable; battery operation operates for 25 to 30 hours
- Automatic shutoff saves battery life
- Built-in overload protection
- Specific gravity and density measurements with built-in below-balance weighing mechanism
- Weighing modes: g, kg, mg, carats, lb., oz., ounces troy, grains, pennyweight, mommes, mesghals, taels (three variants), ticals, tolas and bahts
- Large, high-contrast, seven-segment LCD
- Automatic calibration with external weights
- Large weighing pan supports loading efficiency
- PL-S models offer the option of a built-in rechargeable storage battery
Order Info
Electrical Requirements: 105/120V 50/60Hz. Dimensions: 8.87 x 7.62 x 5.37 in. (22.5 x 19.4 x 13.7cm); except PL303 and PL3002 which are 13.25 x 9.37 x 11.25 (33.5 x 23.8 x 28.7cm). Net Weight: 3.37 lb. (1.5kg); except PL83-S and PL303 which are 12.12 lb. (5.5kg). A variety of accessories are available including optional carrying case, in-use covers, and power adapters.
Warranty and Services
One year
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.