Learn More
Mettler Toledo™ Conductivity Solutions Available on GSA/VA Contract for Federal Government customers only.
Click to view available options
Quantity:
30 x 20mL solution s
30 x 20mL solution
500mL solution
Conductivity:
12.88mS/cm
1413μS/cm
1413uS/cm
84uS/cm
For SevenCompact™ and SevenExcellence™ Benchtop Conductivity Meters and SevenGo™ Portable Conductivity Meters
  • Certified conductivity standards
  • Used to maintain accuracy of conductivity meter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Packaging Bottle -1413μS/cm
Quantity 500mL solution
Conductivity 1413uS/cm
Product Type Mettler Toledo Conductivity Solutions