Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Detachable Cables for S7 Screw Head Electrodes

Connects electrodes with an S7 screw head to meters with specified connector. Mettler Toledo Detachable Cables can be used with meters that have a BNC, 2mm Banana or US Standard connector.

Specifications

Type Cable
Length (English) Cable 47.2 in.
Length (Metric) Cable 1.2m
View More Specs

Products 1
Catalog Number Mfr. No. Connector Types For Use With Price Quantity & Availability  
Catalog Number Mfr. No. Connector Types For Use With Price Quantity & Availability  
01-911-280 View Documents Mettler Toledo™
52300014
US Standard S7 Screw head electrode with US standard connector This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Detachable cables that attach to electrodes with an S7 screw head
  • Length 1.2m (3.9 ft.)

Specifications

Specifications

Cable
47.2 in.
1.2m
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit