Learn More
Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes Available on GSA/VA Contract for Federal Government customers only.
Replacement parts for DL31 Karl Fischer titrators.
Manufacturer:  Mettler Toledo™ 89599
 View more versions of this product
Catalog No. 01-910-725
Add to cart
Description
Description
  • For use with DL31 Karl Fischer titrators
  • Type: Combined Silver Ring Electrode
  • For argentometric titrations
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
DL31 Karl Fischer titrator
0° to 80°C
For argentometric titrations
0° to 176°F
Combined Silver Ring Electrode