Learn More
Mettler Toledo™ Accessories for EasyPlus™ Titrator
For use with Mettler Toledo EasyPlus Titrators.
Supplier: Mettler Toledo™ 30065502
Description
- Vpac Acid
- Potassium hydrogen phthalate (KHP)
- Titrant: NaOH 0.1 mol/L
- For performance verification
Specifications
Acid Titration Standard (Potassium hydrogen phthalate (KHP) |