Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Electrode Storage Vial

For storage of pH electrodes.

Manufacturer:  Mettler Toledo™ 51343320

Catalog No. 01-911-508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Vials keep tips of electrodes wet during storage
  • Holds 12mm (0.47 in.) electrode
  • 12mm (0.47 in.) entryway in vial cap
  • Material: Polyethylene

Specifications

Specifications

Electrode storage vial
50.8mm
Electrode
2 in.
Polyethylene
Storage Vial
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit