Learn More
Mettler Toledo™ METTLER TOLEDO EQPac Comprehensive Qualification, Regulated Balances (IQ/OQ/PQ, USP Certificate)
A professionally executed installation and qualification is essential in achieving the best results from any instrument. METTLER TOLEDO EQPac Comprehensive Qualification for Regulated Balances provide the peace of mind that your instrument is performing perfectly from the beginning.
Manufacturer:  Mettler Toledo™ B39990003
Catalog No. 01-909-402
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
The EQPac for Balances with USP Certificate is designed for highly regulated industries when product safety is of utmost importance and where minimizing the risk of failing an audit is critical.
  • Audit proof documentation used to qualify equipment at every step
  • Documentation of equipment details, setup, maintenance plan, expanded function and performance testing
  • Comprehensive equipment qualification supports testing for process performance
  • Ordering Information: Please note pricing is for Zone 1; please contact your local representative for pricing in Zone 2 regions as additional travel fees may apply
Specifications
Specifications
USP Certificate
Recommended for following industries: Pharma, Biotech, Cosmetics, Food, Hazardous, Medical, Chemical Ingredient Suppliers
Regulated Balances
EQPac Comprehensive Qualification for Regulated Balances (IQ/OQ/PQ, USP Certificate)