Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Excellence Plus XP Analytical Balances

Hands-free operation and unmatched weighing performance

Manufacturer:  Mettler Toledo™ 11106027

 View more versions of this product

Catalog No. 01-911-352

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Analytical balances offer high speed, high performance and security features for measurements, users and data.

SmartGrid™ Weighing Pan

  • Minimizes effects of turbulence in the weighing chamber
  • Ensures more accurate results and shorter stabilization times
  • Smallest sample sizes can be weighed-in according to regulations
  • Increases the yield of expensive substances

SmartSens™ Sensors
  • For hands-free operation and personnel security
  • Enable user to tare, open and close draft shield or print without touching the balance
  • Promotes safe handling of samples, prevention of spills, minimized contamination risk

SmartScreen™ Display
  • Color-guided touchscreen for safe operation
  • Individual, color-coded profiles can be configured and documented for up to eight users or jobs
  • Protects users, data and processes, and guarantees seamless traceability

ErgoClips™
  • Facilitate secure placement and efficient handling of tare containers
  • Farraday effect eliminates negative effects of statically charged tare containers

LevelControl and MinWeigh™
  • For full regulatory compliance
  • LevelControl warns if balance is not correctly leveled
  • MinWeigh signals when sample has not reached minimum weight defined by regulations

Standard Features

  • Prepared for weighing below the balance
  • User administration and password protection
  • Target weighing
  • Statistical evaluation
  • Formulation with security check
  • Density determination
  • Percent weighing
  • Piece counting with reference optimization
  • Alphanumeric data entry of four IDs

 

Description & Specifications

Specifications

Analytical balance
Touch screen
3.07 in.
0.01 / 0.1mg
50/60Hz
23.5cm
48.7cm
50 to 86°F
10 to 30°C
22 lb.
2.87 in.
110/230V 50/60Hz
48.7 x 26.3 x 32.5cm (19 x 10.3 x 12.75 in. )
1 Year
RS232C
32.5cm
XP205DR
10.3 in.
0.8A
GLP, ISO 9001, UL, CSA
7.8cm
9.26 in.
Automatic and detachable draft shield, ProFACT automatic calibration technology, RS232C interface, option slot for second interface (choice of seven different interfaces), inner draft shield plate (adjustable), protective cover, detachable and adjustable terminal
19 in.
1.5 / 2.5 sec.
100g
110/230V
10kg
7.3cm
0.25mg
1.4mg
82 / 220g
12.75 in.
±0.15mg
<±0.015, 0.06mg
26.3cm
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit